Assay Details
Gene Symbol: PAPOLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198601.3 | NP_001185530.1 | 18 - 19 | 2043 | 76 | |
XM_015287288.2 | XP_015142774.1 | 18 - 19 | 2050 | 76 | ||
XM_015287291.2 | XP_015142777.1 | 18 - 19 | 2047 | 76 | ||
GenBank mRNA | AF061758.1 | - | 14 - 15 | 1416 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
