Assay Details
Gene Symbol: RPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277679.1 | NP_001264608.1 | 1 - 2 | 92 | 106 | |
NR_102416.1 | 1 - 2 | 85 | 106 | |||
GenBank mRNA | AJ395262.1 | - | 1 - 2 | 85 | 106 | |
BX934661.2 | - | 1 - 2 | 88 | 106 | ||
CR390036.1 | - | 1 - 2 | 92 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
