Assay Details
Gene Symbol: RPLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_424134.6 | XP_424134.1 | 3 - 4 | 245 | 88 | |
GenBank mRNA | BX932378.2 | - | 2 - 3 | 583 | 88 | |
BX935793.2 | - | 3 - 4 | 222 | 88 | ||
CR406227.1 | - | 3 - 4 | 262 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
