Assay Details
Gene Symbol: RELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015281000.2 | XP_015136486.1 | 9 - 10 | 1017 | 74 | |
XM_025152482.1 | XP_025008250.1 | 2 - 3 | 289 | 74 | ||
XM_025152485.1 | XP_025008253.1 | 10 - 11 | 1070 | 74 | ||
XM_025152486.1 | XP_025008254.1 | 10 - 11 | 1074 | 74 | ||
GenBank mRNA | CR390297.1 | - | 4 - 5 | 487 | 74 | |
CR407478.1 | - | 2 - 3 | 288 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025152482.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
