Assay Details
Gene Symbol: REL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167726.1 | NP_001161198.1 | 8 - 9 | 1029 | 93 | |
XM_015283288.2 | XP_015138774.2 | 8 - 9 | 947 | 93 | ||
GenBank mRNA | M26381.1 | - | 7 - 8 | 908 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
