Assay Details
Gene Symbol: RIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001234688.5 | XP_001234689.4 | 6 - 7 | 490 | 114 | |
GenBank mRNA | BX932754.1 | - | 6 - 7 | 482 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
