Assay Details
Gene Symbol: ATP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199554.1 | NP_001186483.1 | 5 - 6 | 560 | 104 | |
XM_015281404.2 | XP_015136890.1 | 5 - 6 | 572 | 104 | ||
XM_025147802.1 | XP_025003570.1 | 4 - 5 | 600 | 104 | ||
XM_025147804.1 | XP_025003572.1 | 5 - 6 | 633 | 104 | ||
XM_025147808.1 | XP_025003576.1 | 5 - 6 | 519 | 104 | ||
XM_025147815.1 | XP_025003583.1 | 5 - 6 | 620 | 104 | ||
XM_025147817.1 | XP_025003585.1 | 5 - 6 | 526 | 104 | ||
XM_025147823.1 | XP_025003591.1 | 6 - 7 | 636 | 104 | ||
XM_025147830.1 | XP_025003598.1 | 5 - 6 | 656 | 104 | ||
GenBank mRNA | BU400702.1 | - | 3 - 4 | 307 | 104 | |
BU409185.1 | - | 5 - 6 | 588 | 104 | ||
CR352492.1 | - | 5 - 6 | 563 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
