Assay Details
Gene Symbol: OCIAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420717.6 | XP_420717.2 | 4 - 5 | 304 | 66 | |
GenBank mRNA | BX930725.2 | - | 4 - 5 | 303 | 66 | |
BX933413.1 | - | 4 - 5 | 330 | 66 | ||
BX934873.1 | - | 4 - 5 | 352 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
