Assay Details
Gene Symbol: PRTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199585.1 | NP_001186514.1 | 4 - 5 | 407 | 125 | |
XM_015281378.2 | XP_015136864.1 | 4 - 5 | 583 | 125 | ||
GenBank mRNA | BX932120.2 | - | 4 - 5 | 407 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
