Assay Details
Gene Symbol: C1QTNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001232333.5 | XP_001232334.1 | 3 - 4 | 642 | 78 | |
XM_420778.6 | XP_420778.1 | 3 - 4 | 913 | 78 | ||
GenBank mRNA | BX932235.1 | - | 1 - 2 | 174 | 78 | |
BX950284.2 | - | 3 - 4 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
