Assay Details
Gene Symbol: CCNJL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293663.2 | XP_015149149.1 | 4 - 5 | 113 | ||
XM_015293664.2 | XP_015149150.1 | 4 - 5 | 113 | |||
GenBank mRNA | BX950791.1 | - | 4 - 5 | 801 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
