Assay Details
Gene Symbol: G6PC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003641648.4 | XP_003641696.1 | 4 - 5 | 661 | 70 | |
XM_004942769.3 | XP_004942826.1 | 6 - 7 | 831 | 70 | ||
XM_004942770.3 | XP_004942827.1 | 4 - 5 | 660 | 70 | ||
XM_025152633.1 | XP_025008401.1 | 7 - 8 | 916 | 70 | ||
XM_422017.5 | XP_422017.2 | 6 - 7 | 851 | 70 | ||
GenBank mRNA | BX931325.2 | - | 6 - 7 | 805 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
