Assay Details
Gene Symbol: SLC7A6OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293299.1 | NP_001280228.1 | 3 - 4 | 805 | 137 | |
GenBank mRNA | BU247753.1 | - | 2 - 3 | 407 | 137 | |
BX934607.2 | - | 3 - 4 | 808 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
