Assay Details
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288444.2 | XP_015143930.1 | 10 - 11 | 937 | 76 | |
XM_421646.6 | XP_421646.1 | 10 - 11 | 843 | 76 | ||
GenBank mRNA | BX935521.1 | - | 11 - 12 | 845 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
