Assay Details
Gene Symbol: OTUD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004947487.3 | XP_004947544.3 | 3 - 4 | 399 | 71 | |
XM_424363.6 | XP_424363.6 | 3 - 4 | 462 | 71 | ||
GenBank mRNA | BX929705.1 | - | 3 - 4 | 430 | 71 | |
CR524403.1 | - | 3 - 4 | 412 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
