Assay Details

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025148721.1, XM_025148722.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
