Assay Details
Gene Symbol: PTCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001369063.1 | NP_001355992.1 | 4 - 5 | 943 | 148 | |
GenBank mRNA | CR390934.1 | - | 4 - 5 | 545 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() |
