Assay Details
Gene Symbol: BAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419897.6 | XP_419897.5 | 3 | 782 | 141 | |
GenBank mRNA | BX933957.2 | - | 3 | 751 | 141 | |
CR406498.1 | - | 3 | 751 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
