Assay Details
Gene Symbol: IL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204608.1 | NP_989939.1 | 3 - 4 | 216 | 107 | |
XM_015297944.2 | XP_015153430.1 | 4 - 5 | 508 | 107 | ||
XM_015297945.2 | XP_015153431.1 | 3 - 4 | 502 | 107 | ||
XM_015297946.2 | XP_015153432.1 | 5 - 6 | 530 | 107 | ||
XM_015297947.2 | XP_015153433.1 | 4 - 5 | 487 | 107 | ||
XM_015297948.2 | XP_015153434.1 | 4 - 5 | 279 | 107 | ||
XM_025143170.1 | XP_024998938.1 | 4 - 5 | 272 | 107 | ||
GenBank mRNA | AJ276025.1 | - | 2 - 3 | 207 | 107 | |
AJ277865.1 | - | 3 - 4 | 216 | 107 | ||
AY628648.2 | - | 2 - 3 | 207 | 107 | ||
AY775780.1 | - | 3 - 4 | 213 | 107 | ||
AY775781.1 | - | 3 - 4 | 213 | 107 | ||
AY775782.1 | - | 3 - 4 | 213 | 107 | ||
AY920749.1 | - | 2 - 3 | 210 | 107 | ||
EU334505.1 | - | 3 - 4 | 213 | 107 | ||
EU747333.1 | - | 3 - 4 | 213 | 107 | ||
FJ788632.1 | - | 3 - 4 | 213 | 107 | ||
FJ882408.1 | - | 3 - 4 | 214 | 107 | ||
HM854281.1 | - | 3 - 4 | 213 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
