Assay Details
Gene Symbol: OPN4-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204625.3 | NP_989956.3 | 3 - 4 | 478 | 154 | |
XM_015276115.2 | XP_015131601.1 | 13 - 14 | 3304 | 154 | ||
XM_015276116.2 | XP_015131602.1 | 13 - 14 | 8268 | 154 | ||
XM_015276120.2 | XP_015131606.1 | 5 - 6 | 1357 | 154 | ||
XM_025149493.1 | XP_025005261.1 | 13 - 14 | 8342 | 154 | ||
XM_025149494.1 | XP_025005262.1 | 11 - 12 | 8372 | 154 | ||
XM_025149495.1 | XP_025005263.1 | 13 - 14 | 5480 | 154 | ||
XM_025149496.1 | XP_025005264.1 | 12 - 13 | 8346 | 154 | ||
XM_025149497.1 | XP_025005265.1 | 13 - 14 | 8333 | 154 | ||
XM_025149498.1 | XP_025005266.1 | 12 - 13 | 8158 | 154 | ||
GenBank mRNA | AY036061.1 | - | 3 - 4 | 480 | 154 | |
EU124630.1 | - | 3 - 4 | 480 | 154 | ||
EU124631.1 | - | 3 - 4 | 480 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
