Assay Details
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204706.1 | NP_990037.1 | 2 - 3 | 471 | 121 | |
GenBank mRNA | AF275944.1 | - | 2 - 3 | 471 | 121 | |
AF290888.1 | - | 1 - 2 | 282 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
