Assay Details
Gene Symbol: CHRNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204812.2 | NP_990143.2 | 4 - 5 | 365 | 60 | |
XM_015280411.2 | XP_015135897.1 | 4 - 5 | 806 | 60 | ||
XM_025144652.1 | XP_025000420.1 | 3 - 4 | 2238 | 60 | ||
XM_025144653.1 | XP_025000421.1 | 3 - 4 | 2234 | 60 | ||
GenBank mRNA | X83739.2 | - | 365 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
