Assay Details
Gene Symbol: CHRNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204819.1 | NP_990150.1 | 3 - 4 | 397 | 130 | |
GenBank mRNA | J05643.2 | - | 3 - 4 | 397 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
