Assay Details
Gene Symbol: CMTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001778.2 | NP_001001778.2 | 2 - 3 | 300 | 120 | |
GenBank mRNA | AB106870.1 | - | 2 - 3 | 298 | 120 | |
AJ720911.1 | - | 2 - 3 | 300 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
