Assay Details
Gene Symbol: DEGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012565.1 | NP_001012583.1 | 2 - 3 | 869 | 72 | |
GenBank mRNA | AJ851729.1 | - | 2 - 3 | 869 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
