Assay Details
Gene Symbol: TIPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012690.1 | NP_001012708.1 | 4 - 5 | 459 | 76 | |
XM_015292095.2 | XP_015147581.1 | 4 - 5 | 76 | |||
GenBank mRNA | AJ851484.1 | - | 4 - 5 | 459 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
