Assay Details
Gene Symbol: EIF3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012771.2 | NP_001012789.1 | 6 - 7 | 638 | 86 | |
XM_015292113.2 | XP_015147599.1 | 6 - 7 | 639 | 86 | ||
GenBank mRNA | AJ720180.1 | - | 6 - 7 | 639 | 86 | |
CR387918.1 | - | 4 - 5 | 438 | 86 | ||
CR523714.1 | - | 3 - 4 | 363 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
