Assay Details
Gene Symbol: CAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024830.2 | NP_001020001.2 | 3 - 4 | 361 | 74 | |
GenBank mRNA | AY817057.1 | - | 3 - 4 | 361 | 74 | |
DQ092352.1 | - | 3 - 4 | 361 | 74 | ||
FJ938358.1 | - | 3 - 4 | 361 | 74 | ||
HQ640432.1 | - | 4 - 5 | 361 | 74 | ||
KT962965.1 | - | 4 - 5 | 361 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
