Assay Details
Gene Symbol: HSD11B1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001201.1 | NP_001001201.1 | 7 - 8 | 793 | 70 | |
XM_015299681.2 | XP_015155167.2 | 5 - 6 | 1188 | 70 | ||
XM_015299683.2 | XP_015155169.2 | 6 - 7 | 1030 | 70 | ||
XM_015299684.2 | XP_015155170.2 | 6 - 7 | 1026 | 70 | ||
XM_015299685.2 | XP_015155171.2 | 6 - 7 | 1080 | 70 | ||
XM_015299686.2 | XP_015155172.2 | 6 - 7 | 950 | 70 | ||
XM_015299687.2 | XP_015155173.2 | 6 - 7 | 904 | 70 | ||
XM_015299691.2 | XP_015155177.2 | 5 - 6 | 957 | 70 | ||
XM_015299692.1 | XP_015155178.1 | 6 - 7 | 1051 | 70 | ||
XM_015299693.2 | XP_015155179.2 | 6 - 7 | 1023 | 70 | ||
XM_025144260.1 | XP_025000028.1 | 5 - 6 | 1188 | 70 | ||
XM_025144261.1 | XP_025000029.1 | 6 - 7 | 962 | 70 | ||
GenBank mRNA | AY578179.1 | - | 7 - 8 | 793 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
