Assay Details
Gene Symbol: IGFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001294.1 | NP_001001294.1 | 2 - 3 | 666 | 154 | |
GenBank mRNA | AY591922.1 | - | 2 - 3 | 666 | 154 | |
CR407577.1 | - | 2 - 3 | 599 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() |
