Assay Details
Gene Symbol: MPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001782.1 | NP_001001782.1 | 9 - 10 | 1600 | 135 | |
XM_015290974.2 | XP_015146460.1 | 8 - 9 | 2453 | 135 | ||
XM_025152794.1 | XP_025008562.1 | 8 - 9 | 2453 | 135 | ||
XM_025152796.1 | XP_025008564.1 | 8 - 9 | 2444 | 135 | ||
XM_025152797.1 | XP_025008565.1 | 9 - 10 | 2387 | 135 | ||
XM_025152799.1 | XP_025008567.1 | 9 - 10 | 2378 | 135 | ||
XM_025152800.1 | XP_025008568.1 | 9 - 10 | 2369 | 135 | ||
GenBank mRNA | AY613435.1 | - | 9 - 10 | 1600 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
