Assay Details
Gene Symbol: KIF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204975.1 | NP_990306.1 | 10 - 11 | 1405 | 73 | |
XM_015278192.2 | XP_015133678.1 | 10 - 11 | 1462 | 73 | ||
GenBank mRNA | U18309.1 | - | 10 - 11 | 1405 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
