Assay Details
Gene Symbol: KIF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204975.1 | NP_990306.1 | 13 - 14 | 1760 | 124 | |
XM_015278192.2 | XP_015133678.1 | 13 - 14 | 1817 | 124 | ||
GenBank mRNA | U18309.1 | - | 13 - 14 | 1760 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
