Assay Details
Gene Symbol: KIF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204975.1 | NP_990306.1 | 18 - 19 | 2320 | 110 | |
XM_015278192.2 | XP_015133678.1 | 18 - 19 | 2377 | 110 | ||
GenBank mRNA | U04821.1 | - | 1 - 2 | 72 | 110 | |
U18309.1 | - | 18 - 19 | 2320 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
