Assay Details
Gene Symbol: KIF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204975.1 | NP_990306.1 | 25 - 26 | 3164 | 75 | |
XM_015278192.2 | XP_015133678.1 | 25 - 26 | 3221 | 75 | ||
GenBank mRNA | U04821.1 | - | 8 - 9 | 916 | 75 | |
U18309.1 | - | 25 - 26 | 3164 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
