Assay Details
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205017.1 | NP_990348.1 | 5 - 6 | 468 | 147 | |
XM_015294138.2 | XP_015149624.1 | 6 - 7 | 668 | 147 | ||
GenBank mRNA | U79755.1 | - | 5 - 6 | 468 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
