Assay Details
Gene Symbol: CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205317.1 | NP_990648.1 | 3 - 4 | 560 | 93 | |
GenBank mRNA | X04810.1 | - | 336 | 93 | ||
X12639.1 | - | 324 | 93 | |||
Z14957.1 | - | 560 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
