Assay Details
Gene Symbol: PCDH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214672.1 | NP_999837.1 | 3 - 4 | 3046 | 77 | |
XM_015285363.2 | XP_015140849.1 | 3 - 4 | 4895 | 77 | ||
XM_015285364.2 | XP_015140850.1 | 3 - 4 | 4895 | 77 | ||
XM_015285365.2 | XP_015140851.1 | 3 - 4 | 4895 | 77 | ||
XM_015285366.2 | XP_015140852.1 | 3 - 4 | 4895 | 77 | ||
XM_015285367.2 | XP_015140853.1 | 3 - 4 | 4895 | 77 | ||
GenBank mRNA | AF334802.1 | - | 3 - 4 | 3046 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
