Assay Details
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030804.1 | NP_001025975.1 | 5 - 6 | 506 | 86 | |
GenBank mRNA | AJ720842.1 | - | 5 - 6 | 506 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
