Assay Details
Gene Symbol: MLX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030930.1 | NP_001026101.1 | 5 - 6 | 394 | 150 | |
GenBank mRNA | AJ720035.1 | - | 5 - 6 | 394 | 150 | |
BX929898.2 | - | 2 - 3 | 133 | 150 | ||
EF426481.1 | - | 5 - 6 | 381 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
