Assay Details
Gene Symbol: STK17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030995.1 | NP_001026166.1 | 1 - 2 | 272 | 79 | |
GenBank mRNA | AJ719290.1 | - | 1 - 2 | 301 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
