Assay Details
Gene Symbol: CTSO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031129.1 | NP_001026300.1 | 5 - 6 | 703 | 112 | |
XM_015285286.2 | XP_015140772.1 | 5 - 6 | 1279 | 112 | ||
GenBank mRNA | AJ719384.1 | - | 5 - 6 | 703 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
