Assay Details
Gene Symbol: AREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031537.1 | NP_001026708.1 | 2 - 3 | 273 | 67 | |
GenBank mRNA | AY725836.1 | - | 2 - 3 | 273 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
