Assay Details
Gene Symbol: COX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030700.1 | NP_001025871.1 | 5 - 6 | 736 | 96 | |
GenBank mRNA | AJ720296.1 | - | 5 - 6 | 736 | 96 | |
CR523234.1 | - | 4 - 5 | 428 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
