Assay Details
Gene Symbol: NAPEPLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030730.1 | NP_001025901.1 | 1 - 2 | 222 | 95 | |
GenBank mRNA | AJ719820.1 | - | 1 - 2 | 222 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
