Assay Details
Gene Symbol: C1S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030777.1 | NP_001025948.1 | 9 - 10 | 1170 | 71 | |
GenBank mRNA | AJ851617.1 | - | 9 - 10 | 1170 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
