Assay Details
Gene Symbol: GPR89A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030791.1 | NP_001025962.1 | 10 - 11 | 950 | 140 | |
XM_025149429.1 | XP_025005197.1 | 10 - 11 | 1162 | 140 | ||
XM_025149433.1 | XP_025005201.1 | 10 - 11 | 1243 | 140 | ||
GenBank mRNA | AJ851452.1 | - | 10 - 11 | 950 | 140 | |
CR523472.1 | - | 3 - 4 | 300 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
