Assay Details
Gene Symbol: MTSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030954.1 | NP_001026125.1 | 10 - 11 | 1538 | 86 | |
XM_015283097.2 | XP_015138583.1 | 11 - 12 | 1587 | 86 | ||
XM_015283098.2 | XP_015138584.1 | 11 - 12 | 1587 | 86 | ||
XM_015283099.2 | XP_015138585.1 | 10 - 11 | 1575 | 86 | ||
GenBank mRNA | AJ719272.1 | - | 10 - 11 | 1538 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
