Assay Details
Gene Symbol: MTSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030954.1 | NP_001026125.1 | 11 - 12 | 1741 | 82 | |
XM_015283091.2 | XP_015138577.1 | 13 - 14 | 1910 | 82 | ||
XM_015283092.2 | XP_015138578.1 | 13 - 14 | 1910 | 82 | ||
XM_015283093.2 | XP_015138579.1 | 13 - 14 | 1907 | 82 | ||
XM_015283094.2 | XP_015138580.1 | 12 - 13 | 1898 | 82 | ||
XM_015283095.2 | XP_015138581.1 | 13 - 14 | 1826 | 82 | ||
XM_015283097.2 | XP_015138583.1 | 12 - 13 | 1790 | 82 | ||
XM_015283098.2 | XP_015138584.1 | 12 - 13 | 1790 | 82 | ||
XM_015283099.2 | XP_015138585.1 | 11 - 12 | 1778 | 82 | ||
XM_015283100.2 | XP_015138586.1 | 13 - 14 | 1910 | 82 | ||
XM_015283101.2 | XP_015138587.1 | 13 - 14 | 1910 | 82 | ||
XM_025147131.1 | XP_025002899.1 | 13 - 14 | 1437 | 82 | ||
GenBank mRNA | AJ719272.1 | - | 11 - 12 | 1741 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
