Assay Details
Gene Symbol: MTSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030954.1 | NP_001026125.1 | 12 - 13 | 1901 | 64 | |
XM_015283092.2 | XP_015138578.1 | 14 - 15 | 64 | |||
XM_015283098.2 | XP_015138584.1 | 13 - 14 | 64 | |||
XM_015283101.2 | XP_015138587.1 | 14 - 15 | 64 | |||
XM_015283105.2 | XP_015138591.1 | 12 - 13 | 64 | |||
GenBank mRNA | AJ719272.1 | - | 12 - 13 | 1901 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
